Learn More
Podocalyxin (PODXL) (Hematopoietic Stem Cell Marker) Mouse, Clone: PODXL/2185, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 5420-MSM5-P0
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| Podocalyxin (PODXL) (Hematopoietic Stem Cell Marker) | |
| Cell Surface and Cytoplasmic | |
| PODXL/2185 | |
| Unconjugated | |
| Mouse | |
| 165-170kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 5420 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human Podocalyxin (apprx. aa 310-447) | |
| HeLa, Raji, Jurkat cells, Angiosarcoma, Breast, Prostate, Liver, Pancreatic, Kidney | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |