Learn More
STEMCELL Technologies PneumaCult™ Airway Organoid Kit
Descripción
- PneumaCult™ Airway Organoid Kit is a serum- and BPE-free culture medium system for efficient establishment and differentiation of airway organoids derived from human bronchial epithelial cells (HBECs)
- After 2D expansion in PneumaCult™-Ex Plus Medium (#05040), PneumaCult™ Airway Organoid Seeding Medium allows for initiation of 3D organoid culture, then PneumaCult™ Airway Organoid Differentiation Medium is used to obtain morphologically relevant airway organoids
- Airway organoids provide a functional in vitro organotypic culture system for studying the airway epithelium
- When fully differentiated, the organoids exhibit a centralized lumen surrounded by a polarized airway epithelial cell layer, comprised of differentiated cell types, such as ciliated cells and goblet cells
- Applications of airway organoid cultures include modeling lung diseases, drug screening for efficacy and toxicity, and studying the functions of the airway epithelium
Especificaciones
Especificaciones
| Tipo de célula | Airway Cells |
| Para utilizar con (aplicación) | Cell Culture; Differentiation; Expansion; Organoid Culture |
| Cantidad | 1 Kit |