missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12917-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. Plastin 1 (otherwise known as Fimbrin) is a third distinct plastin isoform which is specifically expressed at high levels in the small intestine. The L isoform is expressed only in hemopoietic cell lineages, while the T isoform has been found in all other normal cells of solid tissues that have replicative potential (fibroblasts, endothelial cells, epithelial cells, melanocytes, etc.). The C-terminal 570 amino acids of the T-plastin and L-plastin proteins are 83% identical. It contains a potential calcium-binding site near the N terminus. Alternate splicing results in multiple transcript variants.Especificaciones
| PLS3 | |
| Polyclonal | |
| Unconjugated | |
| PLS3 | |
| Plastin 3, plastin 3 (T isoform), PLS3, T plastin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102866, 5358, 81748 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13797, Q63598, Q99K51 | |
| PLS3 | |
| PLS3 Fusion Protein Ag3574 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Spot an opportunity for improvement?Share a Content Correction