missing translation for 'onlineSavingsMsg'
Learn More

Platinum Standard: Pt @ 10000 μg/mL in 20% HCl, VHG™

Código de producto. p-200131854
Click to view available options
Cantidad:
100 mL
250 mL
500 mL
Tamaño de la unidad:
100 ml
250 ml
500 ml
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17913726

Marca: VHG Labs VHGTPTH250

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Platinum Standard: Pt @ 10000 μg/mL in 20% HCl
TRUSTED_SUSTAINABILITY

Specifications

Nombre del producto químico o material Platinum
CAS 6-4-7440
Métodos ICP/ICP-MS
Nombres de analitos o componentes Platinum
Condiciones de envío Room Temperature
Fórmula molecular Pt
Concentración 10000 μg/mL
Certificaciones/conformidad ISO 17034
Sinónimo Pt
Fórmula InChI InChI=1S/Pt
SMILES [Pt]
Nombre IUPAC platinum
Solvent or Matrix Dilute HCl
Peso molecular (g/mol) 195.08
Cantidad 250 mL
Formula Weight (peso de la fórmula) 194.965 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.