Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A
Código de producto. p-200105184 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/8 in
3/16 in
3/8 in
Diámetro exterior (imperial):
1/2 in
1/4 in
3/16 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
Tamaño de la unidad:
15 metros
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30053184 1/8 in 1/4 in 15 metros
30053181 1/4 in 7/16 in 15 metros
30054183 1/4 in 3/8 in 15 metros
30053005 1/16 in 3/16 in 15 metros
30053899 3/8 in 5/8 in 15 metros
30054065 1/4 in 1/2 in 15 metros
30053651 3/16 in 7/16 in 15 metros
30053154 1/2 in 3/4 in 15 metros
30053638 3/16 in 5/16 in 15 metros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Código de producto. 30053184 Proveedor Cole-Parmer N.º de proveedor 5011956
Solo quedan null
Añadir a la cesta
Edge
Añadir a la cesta
E in., hibits very good resilience, kink resistance, and pressure resistance
Superior optical clarity—easily view your sample. Odorless, noncytoto in., ic, and nonhemolytic. Low-binding surface helps minimize particle and odor absorption. Does not contain BPA, phthalates, PVC, or pero in., ides. Cleanroom packed and double bagged package. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-Phthalate;USP;RoHS
Diámetro interior (imperial) 1/8 in
Diámetro exterior (imperial) 1/4 in
Longitud global (imperial) 50 ft.