Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A
Código de producto. 30053899 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/16 in
1/2 in
1/4 in
1/8 in
3/16 in
3/8 in
Diámetro exterior (imperial):
1/2 in
1/4 in
3/16 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
Tamaño de la unidad:
15 metros
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30053899 3/8 in 5/8 in 15 metros
30053184 1/8 in 1/4 in 15 metros
30053181 1/4 in 7/16 in 15 metros
30054183 1/4 in 3/8 in 15 metros
30053005 1/16 in 3/16 in 15 metros
30054065 1/4 in 1/2 in 15 metros
30053651 3/16 in 7/16 in 15 metros
30053154 1/2 in 3/4 in 15 metros
30053638 3/16 in 5/16 in 15 metros
9 options
Código de producto. 30053899
Marca: Cole-Parmer 5011959
Solo quedan null
Añadir a la cesta
Añadir a la cesta
E in., hibits very good resilience, kink resistance, and pressure resistance
Superior optical clarity—easily view your sample. Odorless, noncytoto in., ic, and nonhemolytic. Low-binding surface helps minimize particle and odor absorption. Does not contain BPA, phthalates, PVC, or pero in., ides. Cleanroom packed and double bagged package. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-Phthalate;USP;RoHS
Diámetro interior (imperial) 3/8 in
Diámetro exterior (imperial) 5/8 in
Longitud global (imperial) 50 ft.