missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PKUMDL WQ 2201

Description
PKUMDL WQ 2201 is a negative allosteric modulator of phosphoglycerate dehydrogenase (PHGDH) (IC50 = 35.7 μM). Displays dose-dependent suppression of cell viability in a range of cancer cell lines. Inhibits tumor growth in a xenograft mouse model.
Specifications
Specifications
| Nombre del producto químico o material | 2-Chloro-4-[5-[[2-[(ethylamino)thioxomethyl]hydrazinylidene]methyl]-2-furanyl]benzoic acid |
| CAS | 592474-91-4 |
| Cantidad | 25 mg |
| Fórmula molecular | C15H14ClN3O3S |
| Formula Weight (peso de la fórmula) | 351.81 |
| Pureza | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?