Learn More
PKM1 Mouse anti-Human, Clone: OTI4E8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF190162
Descripción
Synthetic peptide sequence is LVRASSHSTD LMEAMAMGSV Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PKM1 encodes a protein involved in glycolysis. The encoded protein is a pyruvate kinase that catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate to ADP, generating ATP and pyruvate. This protein has been shown to interact with thyroid hormone and may mediate cellular metabolic effects induced by thyroid hormones. This protein has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Several alternatively spliced transcript variants encoding a few distinct isoforms have been reported.Especificaciones
| PKM1 | |
| Monoclonal | |
| Unconjugated | |
| PKM | |
| PKM1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5315 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P14618 | |
| PKM | |
| Synthetic peptide sequence is corresponding to the region encoded by PKM gene M1 isoform | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.