Learn More
PKC delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16862714 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16862714 150 μL 150 microlitros
16872714 20 μL 20 microlitros
2 options
Código de producto. 16862714
Marca: Proteintech 141881AP150UL
Rabbit Polyclonal Antibody
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.
Especificaciones
Antígeno PKC delta
Aplicaciones Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica PRKCD
N.º de referencia del gen P09215, P28867, Q05655
Alias de gen MAY1, PKC delta, PKCD, PRKCD, Protein kinase C delta type, protein kinase C, delta
Símbolos de los genes Prkcd
Especie del huésped Rabbit
Inmunógeno PKC delta Fusion Protein Ag5473
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 170538, 18753, 5580
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG