Learn More
ratiolab™ Pipet Tips
Click to view available options
Color:
Amarillo
Azul
Transparente
Esterilidad:
Estéril
No estéril
Volumen (métrico):
De 0,1 a 10 μL
De 0,5 a 20 μL
De 1 a 200 μL
De 1 a 250 μL
De 100 a 1000 μL
De 100 a 1200 μL
De 500 a 2500 μL
Envase:
Bolsa
MultiRack
Tamaño de la unidad:
paquete de 1000
paquete de 250
paquete de 600
paquete de 960
Descripción
- Guaranteed dust-free due to special ionization treatment
- High transparency due to high quality surfaces
- Minimal residual wetting due to the controlled quality of raw materials
- High volume accuracy due to precise tip orifice
- Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Especificaciones
Especificaciones
| Volumen (métrico) | De 0,5 a 20 μL |
| Esterilidad | Estéril |
| Color | Transparente |
| Esterilizables en autoclave | Autoclavable |
| Envase | MultiRack |
| Cantidad | 10 x 96 Tips/Pk. |
| Material | Polipropileno |