Learn More
ratiolab™ Pipet Tips
Descripción
- Guaranteed dust-free due to special ionization treatment
- High transparency due to high quality surfaces
- Minimal residual wetting due to the controlled quality of raw materials
- High volume accuracy due to precise tip orifice
- Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Especificaciones
Especificaciones
| Volumen (métrico) | De 100 a 1200 μL |
| Esterilidad | No estéril |
| Color | Azul |
| Esterilizables en autoclave | Autoclavable |
| Envase | MultiRack |
| Cantidad | 10 x 60 Tips/Pk. |
| Material | Polipropileno |