Learn More
ratiolab™ Pipet Tips
Descripción
- Guaranteed dust-free due to special ionization treatment
- High transparency due to high quality surfaces
- Minimal residual wetting due to the controlled quality of raw materials
- High volume accuracy due to precise tip orifice
- Together with specially developed Multirack, these make pipetting much simpler and safer in the most sensitive applications
Especificaciones
Especificaciones
| Volumen (métrico) | De 100 a 1000 μL |
| Esterilidad | No estéril |
| Color | Transparente |
| Esterilizables en autoclave | Autoclavable |
| Envase | Bolsa |
| Cantidad | 1000 Tips |
| Material | Polipropileno |