missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Phosphatidylinositol-4-phosphate-5-kinase (PIPK) synthesizes phosphatidylinositol-4,5-bisphosphate, which regulates various processes including cell proliferation, survival, membrane trafficking and cytoskeletal organization. The PIPK family is divided into type I, type II and type III. Each type of the PIPK family phosphorylates distinct substrates. They contain an activation loop, which determines their enzymatic specificity and subcellular targeting. The phosphatidylinositol-4-phosphate-5-kinase type I consists of three members, PIPK I alpha, beta and gamma, which are characterized by phosphorylating PI4P on the 5-hydroxyl. PIPK I alpha, designated PIPK I beta in mouse, is expressed in brain tissue. PIPK I beta, designated PIPK I alpha in mouse, is also called STM7. PIPK I gamma has two variants produced by alternative splicing which are expressed in lung, brain and kidneys.
Especificaciones
Especificaciones
| Antígeno | PIP5K1C |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | PIP5K1C |
| N.º de referencia del gen | O60331 |
| Alias de gen | KIAA0589, LCCS3, PIP5K GAMMA, PIP5K1 gamma, PIP5K1C, PIP5Kgamma, PIP5KIgamma, PtdIns(4)P 5 kinase 1 gamma |
| Símbolos de los genes | PIP5K1C |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?