Learn More
PIP4K2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. p-7208090 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16816914 20 μL 20 microlitros
16806914 150 μL 150 microlitros
2 options
Código de producto. 16816914
Marca: Proteintech 170771AP20UL
Rabbit Polyclonal Antibody
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Especificaciones
Antígeno PIP4K2C
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica PIP4K2C
N.º de referencia del gen O88370, Q8TBX8, Q91XU3
Alias de gen PI(5)P 4 kinase type II gamma, PIP4K2C, PIP4KII gamma, PIP5K2C
Símbolos de los genes PIP4K2C
Especie del huésped Rabbit
Inmunógeno PIP4K2C Fusion Protein Ag10989
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117150, 140607, 79837
Especies diana Mouse, Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG