missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3AP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5121682
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PIK3AP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
PIK3AP1 (Phosphoinositide-3-Kinase Adaptor Protein 1) is a Protein Coding gene. Diseases associated with PIK3AP1 include Hemangioma Of Lung. Among its related pathways are HIV Life Cycle and PI3K/AKT activation. Gene Ontology (GO) annotations related to this gene include identical protein binding and phosphatidylinositol 3-kinase regulatory subunit binding. An important paralog of this gene is BANK1.
Especificaciones
| PIK3AP1 | |
| Polyclonal | |
| Unconjugated | |
| PIK3AP1 | |
| 1810044J04Rik; B cell adaptor protein; B cell phosphoinositide 3-kinase adaptor; BCAP; B-cell adapter for phosphoinositide 3-kinase; B-cell phosphoinositide 3-kinase adapter protein 1; FLJ35564; Phosphoinositide 3-kinase adapter protein 1; phosphoinositide-3-kinase; phosphoinositide-3-kinase adaptor protein 1; PIK3AP1; RP11-34E5.3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 118788, 294048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6ZUJ8 | |
| PIK3AP1 | |
| Recombinant protein Phosphoinositide-3-Kinase Adaptor Protein 1. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido