Learn More
Invitrogen™ PIGC Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA5114167
Código de producto. 17172024
  • 504.00€ / 100 microlitros
Descripción
Immunogen sequence: AVGAVLFALL LMSISCLCPF YLIRLQLFKE NIHGPWDEAE IKEDLSRFLS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
PIGC
Polyclonal
Unconjugated
PIGC
2.4.1.198; 3110030E07Rik; AW212108; GPI2; phosphatidylinositol glycan anchor biosynthesis class C; phosphatidylinositol glycan anchor biosynthesis, class C; phosphatidylinositol glycan, class C; phosphatidylinositol N-acetylglucosaminyltransferase subunit C; Phosphatidylinositol-glycan biosynthesis class C protein; phosphatidylinositol-glycan biosynthesis, class C protein; PIGC; PIG-C
Rabbit
Affinity chromatography
RUO
5279
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q92535
PIGC
Synthetic peptide directed towards the C terminal region of human PIGC (aa 248-297).
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones