Learn More
Thermo Scientific™ Pastillas inhibidoras de fosfatasas Pierce™
Código de producto. 13424676 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
20
Tamaño de la unidad:
20 tabletas
13424676 20 20 tabletas
1 options
Código de producto. 13424676
Marca: Thermo Scientific™ 88667
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Protect serine, threonine and tyrosine phosphorylation in your sample with these broad-spectrum phosphatase inhibitor tablets.
Thermo Scientific™ Pierce Phosphatase Inhibitor Tablets provide complete cell lysate and biological sample protection from dephosphorylation by a broad spectrum of serine and threonine phosphatases.
Pierce Phosphatase Inhibitor Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.
Highlights:
  • Economical more cost effective than other commercially available formulations
  • Easy-to-use the refrigerator-stable tablets are easily dissolved in buffers and lysis reagents
  • Complete the tablets inhibit acid, alkaline, and protein phosphatases
  • Compatible use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
TRUSTED_SUSTAINABILITY
Especificaciones
Formulación Inhibidores en excipientes patentados en viales de vidrio ámbar
Descripción Minipastillas inhibidoras de fosfatasas Pierce
Formulario Tableta
Suficiente para 200 ml de muestra (10 ml por mini comprimido)
Cantidad 20
Composición Fluoruro sódico, ortovanadato de sodio, pirofosfato sódico, beta-glicerofosfato, aprotinina, bestatina, E-64, leupeptina, EDTA