Learn More
PICALM Mouse anti-Human, Unconjugated, Clone: 1F6G7, Proteintech
Código de producto. 16840266 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16840266 20 μL 20 microlitros
16830266 150 μL 150 microlitros
2 options
Código de producto. 16840266
Marca: Proteintech 675641IG20UL
Mouse Monoclonal Antibody
The PICALM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PICALM. This antibody recognizes human, mouse, rat antigen. The PICALM antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
Antígeno PICALM
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Monoclonal
Clon 1F6G7
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 8301
Especies diana Human, Rat, Mouse
Isotype IgG1