missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHOX2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA529025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, A431, HeLaS3. Predicted reactivity: Mouse (100%), Xenopus laevis (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
Especificaciones
| PHOX2B | |
| Polyclonal | |
| Unconjugated | |
| PHOX2B | |
| dilated pupils 1; Dilp1; GENA 269; LOW QUALITY PROTEIN: paired mesoderm homeobox protein 2B; NBLST2; NBPhox; neuroblastoma paired-type homeobox protein; neuroblastoma Phox; paired like homeobox 2B; paired mesoderm homeobox 2b; paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B; PHOX2B homeodomain protein; Pmx2b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q99453 | |
| PHOX2B | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PHOX2B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido