missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotyrosine Mouse, Clone: E10, Abnova™
Mouse Monoclonal Antibody
Marca: Abnova MAB14229.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mouse monoclonal antibody raised against Phosphotyrosine.
- High specificity
- High reactivity
Especificaciones
| Phosphotyrosine | |
| Monoclonal | |
| Unconjugated | |
| ELISA (0.1 ug/mL) Flow Cytometry (4 ug/mL) Immunocytochemistry (4 ug/mL) Immunofluorescence (4 ug/mL) Immunoprecipitation (1-4 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A purification | |
| RUO | |
| The antibody is specific for proteins containing phosphotyrosine residues and phosphotyrosine peptides. It does not cross-react with phosphoserine, phosphothreonine or nonphosphorylated tyrosine residues. | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| E10 | |
| Mouse monoclonal antibody raised against Phosphotyrosine. | |
| Lyophilized from 0.5 mg/mL PBS, pH 7.4 (0.02% sodium azide). | |
| Phosphotyrosine containing carrier protein. | |
| 100 μg | |
| Primary | |
| Store at -20°C or lower. Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. The reconstituted antibody can be stored for 2-3 weeks at 4°C. For long term storage, aliquot and st |
|
| IgG1 κ, IgG2a κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only (RUO)
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido