Learn More
Invitrogen™ Phospholamban Monoclonal Antibody (2D12)

Descripción
MA3-922 detects phospholamban (PLB) in rabbit, human, pig, sheep, chicken, canine, guinea porcine, rat, and mouse. MA3-922 has been successfully used in Western blot, immunofluorescence, immunohistochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a 25 kDa protein under non-reducing conditions representing the pentameric form of PLB and a 5 kDa protein under reducing conditions representing the monomeric form of PLB in canine cardiac extracts. Immunofluorescence staining of PLB in rat papillary muscle tissue with MA3-922 results in diffuse staining of the network sarcoplasmic reticulum (SR). MA3-922 can also be used to increase the calcium uptake in ventricular SR vesicles analogous to the phosphorylation of PLB following adrenergic stimulation. This antibody is recommended for immunofluorescent staining of tissues. The MA3-922 immunogen is a synthetic peptide corresponding to residues D(2) K V Q Y L T R S A I R R A S T I E M P Q Q A R(25) of canine PLB. This antibody recognizes an epitope between amino acid residues 9-17 of canine PLB.
Especificaciones
Especificaciones
| Antígeno | Phospholamban |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 2D12 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide |
| génica | PLN |
| N.º de referencia del gen | P26677, P26678, P61012, P61013, P61014, P61015, P61016 |
| Alias de gen | cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.