missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TDP43 (Ser409, Ser410) Mouse anti-Human, Mouse, Rat, Clone: 1A2C1, Proteintech
Mouse Monoclonal Antibody
135.00€ - 360.00€
Especificaciones
| Antígeno | Phospho-TDP43 (Ser409, Ser410) |
|---|---|
| Clon | 1A2C1 |
| Concentración | 1.5 mg/mL |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
16828085
|
Proteintech
66318-1-IG-20UL |
20 μL |
135.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
16818085
|
Proteintech
66318-1-IG-100UL |
100 μL |
360.00€
100 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Beschreibung
66318-1-Ig is a phospho-dependent monoclonal antibody specifically recognizing phospho-TDP43 (409/410). It does not react with native TDP-43.
Various biochemical, physiological and behavioural processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/posttranslation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.Spezifikation
| Phospho-TDP43 (Ser409, Ser410) | |
| 1.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ALS10, phospho(409/410)-TDP43, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
| Tardbp | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A2C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q13148, Q921F2 | |
| 230908, 23435, 298648 | |
| Peptide | |
| Primary | |
| -20°C | |
| TARDBP |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts