missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Tau (Thr231) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10707323
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10707323 100 μL cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10707323

Marca: Invitrogen™ 44746G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Phospho-Tau (Thr231) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-Tau (Thr231)
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Formulación Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
génica MAPT
N.º de referencia del gen P10636, P10637
Alias de gen AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287)
Símbolos de los genes MAPT
Especie del huésped Rabbit
Inmunógeno The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Tau that contains threonine 231. The sequence is conserved in mouse and rat.
Método de purificación Antigen Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17762, 4137
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.