Learn More
Invitrogen™ Phospho-Tau (Ser396) Monoclonal Antibody (PHF13.6)

Descripción
This monoclonal antibody recognizes phosphorylation-dependent epitopes of ser-396 on PHF-tau as well as fetal tau, but does not recognize autopsy-derived normal adult tau. Phosphoamino ser-396 is one of the two major immunodominant residues of PHF-tau. PHF 13.6 does not recognize unphosphorylated peptide 390-408 or the same peptide phosphorylated at ser-400, thr-403, or ser-404. PHF 13.6 specifically recognizes the ser-396 phosphorylated peptide and all diphosphorylated peptides containing phosphorylated ser-396. Reactivity was confirmed with human PHF-Tau.
Especificaciones
Especificaciones
| Antígeno | Phospho-Tau (Ser396) |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | PHF13.6 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.1% sodium azide; pH 7.4 |
| génica | MAPT |
| N.º de referencia del gen | P10636, P10637, P19332 |
| Alias de gen | AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.