missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Stat3 (Tyr705) Recombinant Rabbit Monoclonal Antibody (Stat3Y705-B12), PE, Invitrogen™

Código de producto. 17136808 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
10 Tests
100 Tests
Tamaño de la unidad:
paquete de 10
paquete de 100
Código de producto. Cantidad unitSize
17136808 10 Tests paquete de 10
17166488 100 Tests paquete de 100
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17136808

Marca: Invitrogen™ MA537217

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Recombinant Monoclonal Antibody

STAT3 belongs to the family of STAT (signal transducers and activators of transcription) proteins which are phosphorylated by receptor associated kinases, translocate to the nucleus, and act as transcription factors in response to cytokines and growth factors. Coactivators such as CREB-binding protein are required for the transcriptional activation by STAT3. STAT3 can also be activated by Interferon-alpha, Interferon-gamma, EGF, PDGF and IL6. Phosphorylation on tyrosine 705 by JAK1 and JAK2 is essential for STAT3 dimer formation, nuclear translocation, and DNA binding activity. In humans, the STAT3 gene is located on the q arm of chromosome 17. STAT3 has been shown to be activated by IFN-alpha but not IFN-beta. The transcription factors associated with STAT3 are c-Jun and cyclic AMP-responsive enhancer binding protein (CREB). STAT3 mediates the expression of a variety of genes in response to cell stimuli, and thus plays a key role in many cellular processes such as cell growth and apoptosis. The small GTPase Rac1 has been shown to bind and regulate the activity of STAT3 while the PIAS3 protein is a specific inhibitor of STAT3. Three alternatively spliced transcript variants encoding distinct isoforms of STAT3 have been described. Deletion of the STAT3 gene in knock-out mice was lethal at the early embryonic stage.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-Stat3 (Tyr705)
Aplicaciones Flow Cytometry
Clasificación Recombinant Monoclonal
Clon Stat3Y705-B12
Conjugado PE
Formulación PBS with 0.20% BSA and 0.09% sodium azide
génica STAT3
N.º de referencia del gen P40763, P42227
Alias de gen 1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3
Símbolos de los genes STAT3
Especie del huésped Rabbit
Inmunógeno A synthetic phospho-peptide corresponding to residues surrounding Tyr705 of human phospho Stat3.
Método de purificación Protein A/G
Cantidad 10 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20848, 6774
Especies diana Human, Mouse
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.