Learn More
Phospho-RPS6 (Ser240/244) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Tienda Proteintech ProductosDescripción
Ribosomal protein S6 (RPS6), also named as Phosphoprotein NP33, may play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA. Ribosomal protein S6 is the major substrate of protein kinases in eukaryote ribosomes. The phosphorylation is stimulated by growth factors, tumor promoting agents, and mitogens and is dephosphorylated at growth arrest.
Especificaciones
Especificaciones
| Antígeno | Phospho-RPS6 (Ser240/244) |
| Aplicaciones | Western Blot, ELISA |
| Clasificación | Polyclonal |
| Concentración | 800 μg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 6194 |
| Especies diana | Human, Mouse |
| Mostrar más |