missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Prolactin (Ser163) Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA513002
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phospho-Prolactin (Ser163) Polyclonal Antibody for DB
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
Especificaciones
| Phospho-Prolactin (Ser163) | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P01236 | |
| PRL | |
| KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S163 of human PRL. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Dot Blot | |
| Unconjugated | |
| PRL | |
| AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido