missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Progesterone Receptor (Ser190) Monoclonal Antibody (1154)

Código de producto. 11559832
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
11559832 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11559832

Marca: Invitrogen™ MA1413

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA1-413 detects phospho-progesterone receptor (pSer190) from human samples. MA1-413 has been successfully used in Western blot, immunohistochemistry, ELISA, immunofluorescence, and immunoprecipitation procedures. By Western blot, this antibody detects ~81 and ~116 kDa proteins representing phospho-progesterone receptor (pSer190) A and B from T47D cell and human breast carcinoma extracts. Immunohistochemical staining of phospho-progesterone receptor (pSer190) (paraffin samples) can be performed using MA1-413. The MA1-413 immunogen is a synthetic peptide corresponding to residues V(184) L P R G L S(p) P A R Q L L(196) of human Progesterone Receptor.

The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-Progesterone Receptor (Ser190)
Aplicaciones ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon 1154
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica PGR
N.º de referencia del gen P06401
Alias de gen 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Símbolos de los genes PGR
Especie del huésped Mouse
Inmunógeno Synthetic phosphopeptide corresponding to residues V(184) L P R G L S(p) P A R Q L L(196) of human Progesterone Receptor.
Método de purificación purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5241
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.