missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PLCg2 (Tyr759) Recombinant Rabbit Monoclonal Antibody (PLCG2Y759-G3), PE, Invitrogen™
Tienda Thermo Scientific ProductosDescripción
Phosphoinositide-specific phospholipase C (PLC) plays a crucial role in the initiation of receptor mediated signal transduction through the generation of the two second messengers, inositol 1,4,5-triphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. There are many mammalian PLC isozymes, including PLC beta1, PLC beta2, PLC beta3, PLC beta4, PLC gamma1, PLC gamma2, PLC delta1, PLC delta2 and PLCe. PLC delta exists as four different isoforms. PLC delta1, a calcium signal amplifier, is activated by an atypical GTP-binding protein. In addition, PLC delta1 is an effector for GTP-binding protein transglutaminase II-mediated oxytocin receptor and alpha1B-adrenoreceptor signaling. Mouse PLC delta1 is highly expressed in brain, heart, lung and testis. PLC delta is abnormally accumulated in autopsied brains with Alzheimer's disease (AD), suggesting that it may play a role in the pathology of AD. PLC delta2 is markedly expressed in type II intestinal metaplasia and in the adenocarcinoma. When PLC delta2 is expressed in type I intestinal metaplasia, the metaplasia is generally considered benignant, yet evolves toward neoplastic transformation. Thus, PLC delta2 expression may be a possible marker of gastric malignant transformation.
Especificaciones
Especificaciones
| Antígeno | Phospho-PLCg2 (Tyr759) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Recombinant Monoclonal |
| Clon | PLCG2Y759-G3 |
| Conjugado | PE |
| Formulación | PBS with 0.20% BSA and 0.09% sodium azide |
| génica | Plcg2 |
| N.º de referencia del gen | P16885, Q8CIH5 |
| Alias de gen | 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2; APLAID; FCAS3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-2; phospholipase C gamma 2; phospholipase C, gamma 2; phospholipase C, gamma 2 (phosphatidylinositol-specific); phospholipase C-gamma-2; Phospholipase C-IV; PLC gamma 2; PLC gamma2; PLC- gamma2; PLC gamma-2; PLCG2; Plcg-2; PLC-gamma; PLCgamma2; PLC-gamma-2; PLC-IV |
| Símbolos de los genes | Plcg2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?