missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PIM1 (Tyr309) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA537683
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control for Western blot is HUVEC cells.
The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
Especificaciones
| Phospho-PIM1 (Tyr309) | |
| Polyclonal | |
| Unconjugated | |
| PIM1 | |
| non-specific serine/threonine protein kinase; Oncogene PIM1; PIM; Pim1; Pim-1; pim-1 kinase 44 kDa isoform; pim-1 oncogene; pim-1 oncogene (proviral integration site 1); Pim-1 proto-oncogene, serine/threonine kinase; proto-oncogene serine/threonine-protein kinase Pim-1; proviral integration site 1; serine/threonine-protein kinase pim-1; serine-threonine kinase PIM-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18712, 24649, 5292 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P06803, P11309, P26794 | |
| PIM1 | |
| Peptide sequence around phosphorylation site of tyrosine 309(H-R-Y(p)-H-G) derived from Human Pim-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido