missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PI3K p85/p55 (Tyr458, Tyr199) Recombinant Rabbit Monoclonal Antibody (PI3KY458-1A11), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA536956
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phosphoinositide 3-kinase (PI3K, phospho PI3K) Binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phospho PI3K is necessary for the insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. During insulin stimulation, it also binds to IRS-1. Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration, and cell survival.
Especificaciones
| Phospho-PI3K p85/p55 (Tyr458, Tyr199) | |
| Recombinant Monoclonal | |
| APC | |
| PIK3R1 | |
| AA414954; AGM7; GRB1; IMD36; mitochondrial poly(ADP-ribose) glycohydrolase; MPPH; MPPH1; OTTHUMP00000009783; OTTHUMP00000009784; p50alpha; p55; p55alpha; p55-GAMMA; p55PIK; p85; p85alpha; p85-ALPHA; P85B; p85beta; p85-BETA; PARG; PARG99; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; phosphatidylinositol 3-kinase regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit beta; phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphoinositide 3-kinase p85 (other splicing variants: p55 and p50); Phosphoinositide 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit 1; phosphoinositide-3-kinase regulatory subunit 2; phosphoinositide-3-kinase regulatory subunit 3; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3K; PI3K regulatory subunit alpha; PI3K regulatory subunit beta; PI3K regulatory subunit gamma; PI3KA; PI3-kinase p85 subunit alpha; PI3-kinase p85 subunit beta; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit beta; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; PI3-kinase subunit p85-beta; Pik3r1; PIK3R2; Pik3r3; poly (ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase; poly(ADP-ribose) glycohydrolase 60 kDa isoform; polypeptide 3; ptdIns-3-kinase p85-alpha; ptdIns-3-kinase p85-beta; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit alpha; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-alpha; ptdIns-3-kinase regulatory subunit p85-beta; RP4-533D7.2 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 18708, 18709, 18710, 5295, 5296, 8503 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| PI3KY458-1A11 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| O00459, O08908, P26450, P27986, Q64143, Q92569 | |
| PIK3R1, Pik3r2, PIK3R3 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr458 of human phospho PI3K p85. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido