Learn More
Phospho-P53 (Ser15) Mouse, anti-Human, Unconjugated, Clone: 1H6G1, Proteintech
Código de producto. 17251476 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17251476 20 μL 20 microlitros
17241476 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17251476 Proveedor Proteintech N.º de proveedor 678261IG20UL
Mouse Monoclonal Antibody
TP53, also known as P53 and NY-CO-13, belongs to the p53 family and has 9 isoforms. In SDS-Page, the observed molecular weight is about 53 kDa. TP53 acts as a tumor suppressor in many tumor types, including growth arrest or apoptosis depending on the physiological circumstances and cell types. It is involved in cell cycle regulation as a trans-activator that acts to negatively regulate cell division by controlling a set of genes required for this process. TP53 Localizes in the nucleus in most cells but found in the cytoplasm in some cells. (PMID: 26166714; PMID: 25225161)
Especificaciones
Antígeno Phospho-P53 (Ser15)
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 1H6G1
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 7157
Especies diana Human