missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MYL9 (Thr18, Ser19) Polyclonal Antibody
GREENER_CHOICE

Produktkod. 11333222
Change view
Klicka för att se tillgängliga alternativ
Cantidad:
100 μL
Förpackningsstorlek:
100 microlitros
Produktkod. Cantidad unitSize
11333222 100 μL 100 microlitros
1 options
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11333222

Brand: Invitrogen™ PA517727

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the cardiac isoform of myosin light chain 2.

Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifikationer

Antígeno Phospho-MYL9 (Thr18, Ser19)
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 20 μg/mL
Conjugado Unconjugated
Formulación 0.01M HEPES with 50% glycerol, 100 μg/mL BSA, 0.15M NaCl and no preservative; pH 7.5
génica MYL9
N.º de referencia del gen P24844, Q9CQ19
Alias de gen 20 kDa myosin light chain; AI327049; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; myl9; myl9.L; myl9b; myl9l; Mylc2c; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light polypeptide 9; myosin regulatory light polypeptide 9b; myosin RLC; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; RLC-C; smooth muscle major isoform; unnamed protein product; XELAEV_18003093mg; zgc:77916
Símbolos de los genes MYL9
Especie del huésped Rabbit
Inmunógeno Synthetic phosphopeptide corresponding to residues surrounding pThr18/Ser19 of human myosin light chain 2.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10398, 98932
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.