Learn More
Phospho-MEK1 (Ser298) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68047-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Phospho-MEK1 (Ser298) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-MEK1 (Ser298). This antibody recognizes human antigen. The Phospho-MEK1 (Ser298) antibody has been validated for the following applications: WB, ELISA analysis.Especificaciones
| MEK1 (Ser298) | |
| Monoclonal | |
| 1000 μg/mL | |
| MAPKK1; MEK1; MKK1; PRKMK1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 40-50 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3F10G10 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5604 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |