missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-LPIN1 (Thr14) Polyclonal Antibody

Description
Phospho-LPIN1 (Thr14) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antígeno | Phospho-LPIN1 (Thr14) |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | LPIN1 |
| N.º de referencia del gen | Q14693, Q91ZP3 |
| Alias de gen | 4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1 |
| Símbolos de los genes | LPIN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?