Learn More
Invitrogen™ Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody

Descripción
PA1-9593 detects Phospho-JNK/SAPK in human, mouse, and rat samples. PA1-9593 has been successfully used in Western blot, immunohistochemistry, and immunocytochemistry procedures. By Western blot, PA1-9593 detects a 46 kDa and 54 kDa protein representing Phospho-JNK/SAPK. PA1-9593 immunogen is a phosphorylated peptide (pT183-P-pY185) based on human JNK.
Especificaciones
Especificaciones
| Antígeno | Phospho-JNK1/JNK2 (Thr183, Tyr185) |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | MAPK8 |
| N.º de referencia del gen | P45983, P45984, P49185, P49186, Q91Y86, Q9WTU6 |
| Alias de gen | AI849689; AI851083; C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; Mapk8; Mapk9; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; OTTHUMP00000019552; p54 gamma; p54a; p54-alpha; p54aSAPK; Phospho-JNK; PRKM; PRKM8; Prkm9; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK; SAPK 1; SAPK gamma; SAPK1; SAPK1A; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1c; stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2 |
| Símbolos de los genes | MAPK8, MAPK9 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.