Learn More
Phospho-ERK1/2 (Thr202/Tyr204) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16826235 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
16826235 20 μL 20 microlitros
16816235 100 μL 100 microlitros
2 options
Código de producto. 16826235
Marca: Proteintech 287331AP20UL
Rabbit Polyclonal Antibody
The Phospho-ERK1/2 (Thr202/Tyr204) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-ERK1/2 (Thr202/Tyr204). This antibody recognizes human, mouse, rat antigen. The Phospho-ERK1/2 (Thr202/Tyr204) antibody has been validated for the following applications: IP, WB, ELISA analysis.
Especificaciones
Antígeno Phospho-ERK1/2 (Thr202/Tyr204)
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Polyclonal
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 5595
Especies diana Human, Rat, Mouse
Isotype IgG