Learn More
Phospho-ERK1/2 (Thr202/Tyr204) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 16816235 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16816235 100 μL 100 microlitros
16826235 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16816235 Proveedor Proteintech N.º de proveedor 287331AP100UL
Rabbit Polyclonal Antibody
The Phospho-ERK1/2 (Thr202/Tyr204) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-ERK1/2 (Thr202/Tyr204). This antibody recognizes human, mouse, rat antigen. The Phospho-ERK1/2 (Thr202/Tyr204) antibody has been validated for the following applications: IP, WB, ELISA analysis.
Especificaciones
Antígeno Phospho-ERK1/2 (Thr202/Tyr204)
Aplicaciones Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Clasificación Polyclonal
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 5595
Especies diana Human, Rat, Mouse
Isotype IgG