missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EEF2 (Thr56) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA597417
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: TRFTD; Positive Samples: NIH/3T3, C6; Cellular Location: Cytoplasm, Nucleus.
Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
Especificaciones
| Phospho-EEF2 (Thr56) | |
| Polyclonal | |
| Unconjugated | |
| EEF2 | |
| EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13629, 1938, 29565 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.50 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P05197, P13639, P58252 | |
| EEF2 | |
| A synthetic phosphorylated peptide around T56 of human Phospho-eEF2-T56 (NP_0019521). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido