missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Claudin 4 (Tyr208) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA564663
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Phospho-Claudin 4 (Tyr208) Polyclonal Antibody detects endogenous levels of Claudin 4 only when phosphorylated at Tyr208.
Claudin-4 belongs to the 20-member family of claudins, transmembrane cellular adhesion proteins localized at epithelial and endothelial tight junctions. Claudins demonstrate distinct, tissue-specific expression patterns, and are essential components of tight junction charge-specific channels that regulate paracellular ion flux. Claudin-4 is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life. The overexpression of Claudin-4 has been shown to decrease paracellular electrical conductance due to a selective decrease in Na+ permeability, with no significant decrease in Cl- permeability. In ovarian carcinoma cells, Claudin-3 and Claudin-4 exhibit upregulation at both mRNA and protein levels. Claudin-4 is also overexpressed in pancreatic cancer tissues and cell lines, and in other epithelial cancers. The Claudin-4 gene is deleted in Williams-Beuren syndrome, a neurodevelopmental disorder affecting multiple systems.
Especificaciones
| Phospho-Claudin 4 (Tyr208) | |
| Polyclonal | |
| Unconjugated | |
| Cldn4 | |
| Cep-r; claudin 4; CLAUDIN4; claudin-4; CLDN4; Clostridium perfringens enterotoxin receptor; Clostridium perfringens enterotoxin receptor 1; CPER; CPE-R; CPE-receptor; Cpetr; CPETR1; h CPE-receptor; hCPE-R; WBSCR8; Williams-Beuren syndrome chromosomal region 8 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1364 | |
| -20°C | |
| Liquid |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| O14493 | |
| Cldn4 | |
| A synthesized peptide derived from human Claudin 4 (Phospho-Tyr208). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido