missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (chk1S317-F10), FITC, Invitrogen™
Tienda Thermo Scientific ProductosDescripción
CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.
Especificaciones
Especificaciones
| Antígeno | Phospho-Chk1 (Ser317) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Recombinant Monoclonal |
| Clon | chk1S317-F10 |
| Conjugado | FITC |
| Formulación | PBS with 0.20% BSA and 0.09% sodium azide |
| génica | Chek1 |
| N.º de referencia del gen | O14757, O35280 |
| Alias de gen | C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W |
| Símbolos de los genes | Chek1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?