missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-CaMKII alpha (Thr286) Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10466223
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10466223 100 μL cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10466223

Marca: Invitrogen™ 44674G

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Phospho-CaMKII alpha (Thr286) Polyclonal Antibody for Western Blot

CAMK2A (CaMKII alpha) is a serine/threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Phospho-CaMKII alpha (Thr286)
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Formulación Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
génica CAMK2A
N.º de referencia del gen P11275, Q9UQM7
Alias de gen alpha CaM kinase II; alpha-CaMKII; Ca2+/calmodulin-dependent protein kinase II; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium/calmodulin dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase II alpha-B subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; calcium/calmodulin-dependent protein kinase type II subunit alpha; CaM kinase II alpha subunit; caM kinase II subunit alpha; CaMK II; CAMK2; Camk2a; CAMKA; CaMKII; CaMK-II alpha subunit; caMK-II subunit alpha; CaMKIINalpha; CaM-kinase II alpha chain; EC 2.7.11.17; KIAA0968; mKIAA0968; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; PK2CDD; PKCCD; R74975
Símbolos de los genes CAMK2A
Especie del huésped Rabbit
Inmunógeno The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human CaMKIIa that contains threonine 286. The sequence is conserved in mouse and rat.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 25400, 815
Especies diana Human, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.