Learn More
Invitrogen™ Phospho-c-Jun (Ser73) Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA517879
Código de producto. 11587120
  • 524.00€ / 100 microlitros
Descripción
It is not recommended to aliquot this antibody.
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Phospho-c-Jun (Ser73)
Polyclonal
Unconjugated
JUN
Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Rabbit
Antigen affinity chromatography, Protein A
RUO
16476, 24516, 3725
-20°C
Liquid
Western Blot, Immunocytochemistry
8 μg/mL
0.01M HEPES with 50% glycerol, 0.15M NaCl, 100μg/mL BSA and no preservative; pH 7.5
P05412, P05627, P17325
JUN
Synthetic phosphopeptide corresponding to residues around pSer73 of human c-Jun.
100 μL
Primary
Human, Mouse, Monkey, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones