missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-c-Cbl (Tyr700) Recombinant Rabbit Monoclonal Antibody (CblY700-E1), Invitrogen™

Código de producto. 17176938 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
20 μL
200 μL
Tamaño de la unidad:
20 microlitros
200 microlitros
Código de producto. Cantidad unitSize
17176938 20 μL 20 microlitros
17116698 200 μL 200 microlitros
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 17176938

Marca: Invitrogen™ MA537351

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Recombinant Monoclonal Antibody

Cbl proteins are a family of ubiquitin protein ligases (E3s) that negatively regulate signaling by targeting activated tyrosine kinases for degradation. Cbl- c is the most recently cloned member of the Cbl proteins and is expressed only in epithelial cells (the other Cbl proteins are ubiquitously expressed). Cbl-c, like the other mammalian Cbl proteins, can ubiquitinate the activated EGFR and target it for degradation. Through interactions with proteins containing SRC homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling.
TRUSTED_SUSTAINABILITY

Specifica

Antígeno Phospho-c-Cbl (Tyr700)
Aplicaciones Flow Cytometry
Clasificación Recombinant Monoclonal
Clon CblY700-E1
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.10% BSA, 50% glycerol and 0.02% sodium azide
génica Cbl
N.º de referencia del gen P22681, P22682
Alias de gen 4732447J05Rik; Cas-Br-M (murine) ecotropic retroviral transforming sequence; Casitas B-lineage lymphoma; casitas B-lineage lymphoma proto-oncogene; CBL; Cbl proto-oncogene; Cbl proto-oncogene, E3 ubiquitin protein ligase; CBL2; Cbl-2; CBL-3; CBLC; c-Cbl; E3 ubiquitin-protein ligase CBL; FRA11B; fragile site, folic acid type, rare, fra(11)(q23.3); NSLL; oncogene CBL2; proto-oncogene c-Cbl; RGD1561386; RING finger protein 55; RING-type E3 ubiquitin transferase CBL; RNF55; RNF57; signal transduction protein CBL
Símbolos de los genes Cbl
Especie del huésped Rabbit
Inmunógeno A synthetic phospho-peptide corresponding to residues surrounding Tyr700 of human phospho c-Cbl.
Método de purificación Protein A/G
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12402, 500985, 867
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG κ
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.