missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phleum pratense Pollen allergen Phl p 2 Polyclonal Antibody
Tienda Thermo Scientific Productos

Description
This antibody has been tested in direct-ELISA.
Phl p 2 is a major respiratory pollen allergen present in Timothy grass (Phleum pratense), which is being recognized by more than 200 million people suffering from allergy worldwide (1). Phl p 2 is a representative of the large family of cross-reacting grass pollen allergens classified as grass allergens group 2/3.
Specifications
Specifications
| Antígeno | Phleum pratense Pollen allergen Phl p 2 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 3.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Alias de gen | allergen Phl p 2; Allergen Phl p II; PHLPII |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant Phleum pratense Pollen allergen Phl p 2 protein (27-122aa). |
| Método de purificación | Protein G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?