missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TSSC3 (Tumor-suppressing STF cDNA 3 protein), also known as PHLDA2 (pleckstrin homology-like domain family A member 2), BWR1C, HLDA2 or IPL, is a cytoplasmic protein that is involved in fetal and placental growth. Expressed at high levels in placenta and adult prostate and at low levels in liver, lung and brain, TSSC3 is an apoptosis-related protein that acts as a negative growth regulator and is expressed during normal human development. TSSC3, a protein expressed from the maternal allele, is imprinted on placenta, liver and fetal tissues during embryogenesis and is removed once development is complete. Defects or alterations in the gene encoding TSSC3 are associated with several afflictions such as lung, ovarian and breast cancer, rhabdomyosarcoma, Beckwith-Wiedemann syndrome, Wilms tumor, low birth weight and adrenocortical carcinoma.
Especificaciones
Especificaciones
| Antígeno | PHLDA2 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.39 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | PHLDA2 |
| N.º de referencia del gen | Q53GA4 |
| Alias de gen | BRW1C, BWR1C, HLDA2, IPL, p17 BWR1C, PHLDA2, TSSC3 |
| Símbolos de los genes | PHLDA2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?