missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGP9.5 Polyclonal Antibody, Invitrogen™
Descripción
PA5-16825 targets Protein Gene Product 9.5 in IHC (P) applications and shows reactivity with Human samples. The PA5-16825 immunogen is recombit full-length human PGP95 protein.
PGP9.5 (Protein gene product 9.5, UCH-L1, PARK5) is a neuron specific protein, structurally and immunologically distinct from neuron specific enolase. PGP9.5 has a molecular weight of 27 kDa and was first identified by high resolution two dimensional PAGE. PGP9.5 is a member of ubiquitin carboxyl-terminal hydrolase family 1 (peptidase family C12) with a ubiquitin carboxyl-terminal hydrolase domain. PGP9.5 is well known for having ubiquitin hydrolase and ligase activities that hydrolyzes small C-terminal adducts of ubiquitin to generate ubiquitin monomers. PGP9.5 is present in neurons and nerve fibers at all levels of the central and peripheral nervous system, in neuroendocrine cells, in segments of the renal tubules, in spermatogonia and Leydig cells of the testis, in ova and in some cells of both the pregt and non-pregt corpus luteum. Over expression of PGP9.5 leads to non-small cell lung cancer while decreased expression leads to Huntington disease and Alzheimer disease. Since PGP9.5 is present in cellular inclusions, it can be a useful as a neuronal marker and in the studies of neurodegenerative disorders such as with Parkinson disease.
Especificaciones
Especificaciones
| Antígeno | PGP9.5 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA and <0.1% sodium azide; pH 7.6 |
| génica | UCHL1 |
| N.º de referencia del gen | P09936, Q00981 |
| Alias de gen | neuron cytoplasmic protein 9.5, ubiquitin C-terminal hydrolase, ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin thioesterase L1 |
| Símbolos de los genes | UCHL1 |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?