missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA544559
This item is not returnable.
View return policy
Description
Peptide sequence: TVEKADNFEY SDPVDGSISR NQGLRLIFTD GSRIVFRLSG TGSAGATIRL Sequence homology: Cow: 93%; Dog: 93%; Goat: 85%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
Specifications
| PGM1 | |
| Polyclonal | |
| Unconjugated | |
| PGM1 | |
| 3230402E02Rik; Aciculin; CDG1T; Glucose phosphomutase 1; glucose phosphomutase 2; GSD14; PGM; PGM 1; PGM 2; Pgm1; Pgm-1; Pgm2; PGM5; PGMRP; PGM-RP; Phosphodeoxyribomutase; phosphoglucomutase 1; phosphoglucomutase 2; phosphoglucomutase 5; phosphoglucomutase isoform 2; phosphoglucomutase isoform1; phosphoglucomutase-1; Phosphoglucomutase-2; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; phosphopentomutase; zgc:63718 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5236 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P36871 | |
| PGM1 | |
| Synthetic peptide directed towards the middle region of human PGM1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction