Learn More
Invitrogen™ PFAS Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA554568
Descripción
Immunogen sequence: LFGCPLLLDD VARESWLLPG SNDLLLEVGP RLNFSTPTST NIVSVCRATG LGPVDRVETT RRYRLSFAHP PSAEVEAIAL ATL Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 84%.
Purines are necessary for many cellular processes, including DNA replication, transcription, and energy metabolism. Ten enzymatic steps are required to synthesize inosine monophosphate (IMP) in the de novo pathway of purine biosynthesis. The enzyme encoded by this gene catalyzes the fourth step of IMP biosynthesis.
Especificaciones
| PFAS | |
| Polyclonal | |
| Unconjugated | |
| PFAS | |
| 4432409B16Rik; FGAM synthase; FGAM synthetase; FGAMS; FGAR amidotransferase; FGARAT; FGAR-AT; Formylglycinamide ribonucleotide amidotransferase; formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Gm18; Kiaa0361; PFAS; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase); pur4; PURL; Sofa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5198 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15067 | |
| PFAS | |
| Recombinant protein corresponding to Human PFAS. Recombinant protein control fragment (Product #RP-93471). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.