missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-06282999, MedChemExpress
Description
PF-06282999 is a potent and selective myeloperoxidase inhibitor which is potential useful for the treatment of cardiovascular diseases.
Specifications
Specifications
| Nombre del producto químico o material | PF-06282999 |
| CAS | 1435467-37-0 |
| Color | Light Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C13H12ClN3O3S |
| Cantidad | 50 mg |
| Información de solubilidad | DMSO : ≥ 32 mg/mL (98.23 mM) |
| SMILES | O=C(N)CN(C(C1=CC(Cl)=CC=C1OC)=C2)C(NC2=O)=S |
| Peso molecular (g/mol) | 325.77 |
| Formula Weight (peso de la fórmula) | 325.77 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?